Learn More
StatLab MasterTech Modified Mayer's Hematoxylin
Click to view available options
:
Pint
Description
- A light delicate hematoxylin with no mucin affinity
- Stain requires no differentiating and no bluing step to achieve optimal results
- Ready-to-use, no filtering is required
Specifications
Specifications
Type | Hematoxylin |
Product Type | Modified Mayer's Hematoxylin |