Learn More
StatLab MasterTech One Step Trichrome Stain (Blue and Red)
Click to view available options
Quantity:
3.5 oz.
1 gal.
Description
- This staining method allows pathologists and researchers to visualize tissue architecture with remarkable clarity, making it invaluable for identifying pathological conditions such as liver fibrosis, myocardial fibrosis, and tumor invasion patterns in cancer research
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Type | Trichrome One-Step Red |
| Product Type | Special Stain Kit Component |
| Quantity | 1 gal. |