Learn More
        Azer Scientific Presto Slide Printer
Description
Our microscope slide printer is an ultraviolet laser model that can integrate into any laboratory or hospital information system. Permanent and resistant to degradation, the laser slide printer offers a high resolution of 500 x 500 dpi and high speed, less than 3 seconds for on-demand printing, or up to 20 slides per minute. Its input cartridge holds up to 100 slides and its output holds up to 35 slides. The on-demand printing model captures detail via barcoding, QR code, logo, alphanumeric and graphic printing. Implementing the new Azer Scientific slide printer aids in a time management workflow that you will not want to resist
Features:
Ultraviolet laser
Permanent and resistant to degradation
Integrate to LIS/HIS system- High resolution; 500 x 500 dpi
High speed; less than 3 seconds for on-demand printing, or up to 20 slides per minute
Input cartridge holds up to 100 slides and output holds up to 35 slides
Continuous and on-demanding printing model
Barcode, QR code, logo, alphanumeric, graphic printing
Specifications
Specifications
| Product Type | Slide Printer | 
| Voltage | 100/115/220/230 V | 
Your input is important to us. Please complete this form to provide feedback related to the content on this product.