Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

General Data Healthcare™ SHUR/Stain™ Manual Stainer Accessories
SDP

Catalog No. 22048005 Shop All Triangle Biomedical Sciences Inc. (TBS) Products
Click to view available options
Product Type:
Digital Timer
Replacement Label Strip, for SS-12L
Replacement Label Strip, for SS-6L
Replacement Label Strip,for SS-3L
Replacement Well, Green
Replacement Well, White
Slide Rack

Accessories for manual stainers (SS-3L, SS-6L, SS-12L)

  • Accessory

Specifications

Product Type Slide Rack
Color Gray
Material Plastic
For Use With (Equipment) SHUR/Stain Manual Stainer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.