Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Butanediol diglycidyl ether

Catalog No. 22089250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
50 g
250 g

Catalog No. 22-089-250G

Supplier: Sigma Aldrich 22089250G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Butanediol diglycidyl ether (BDDE) is a viscous liquid , which is hygroscopic in nature. BDDE is the most commonly used homobifunctional epoxide compound. BDDE may react with amines, hydroxyls, sulfhydryl groups to produce secondary amines, ether or thioether bonds respectively.BDDE is a cross linking agent used for preparing amylose, xylan and hydroxyethyl-celluloseto cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cellsto cross link hyaluronic acid into hydrogelsfor the activation of soluble dextran polymers.Amine or hydroxyl containing bis-epoxy supports are reacted with BDDE to form particles containing terminal epoxide group for immobilization reactions.

Specifications

CAS 2425-79-8
Density 1.1 g/mL (at 25°C)
Boiling Point 266°C
Molecular Formula C10H18O4
Refractive Index n20/D 1.453 (literature)
Linear Formula C10H18O4
Quantity 50 g
MDL Number MFCD00005146
Synonym 1,4-Bis(2,3-epoxypropyloxy)butane
Molecular Weight (g/mol) 202.25
Percent Purity ≥95%
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.