Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Neg-50™ Frozen Section Medium
Shop All Epredia Products
Click to view available options
Quantity:
4 oz. per bottle
4 oz. per bottle, 2/cs.
4 oz. per bottle, 6/cs.
4 oz.per bottle
Color:
Blue
Clear
Five Colors plus Clear
Green
Orange
Pink
Yellow
Description
Yield high-quality sections down to -50°C without chatter using Epredia; Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
- Buffered to ensure proper alkalinity (pH = 8.0); leaves no residue after staining
- Specially formulated to prevent tissue separation from the slide
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.
Specifications
Specifications
| Color | Clear |
| Packaging Type | 2/Cs. |
| Quantity | 4 oz. per bottle, 2/cs. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction