Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MAS™ UA Controls
Click to view available options
Description:
UA Controls, Assayed
Level:
1
2 (abnormal)
Quantity:
15 mL
60 mL
Description
Monitor urine dipstick and microscopic components with Thermo Scientific™ MAS™ UA Controls, bi-level controls with improved room temperature storage capability. Liquid and ready-to-use, this urine control is assayed for most major reagents strips and strip systems. It includes 10 commonly-assayed dipstick tests plus Microalbumin, Creatinine and hCG.
- Level 2 (abnormal)
- Available in convenient 15mL dropper bottles
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | UA Controls, Assayed |
| Detectable Analytes | Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, Osmolality, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells |
| Sample Type | Urine |
| Type | Control |
| Level | 2 (abnormal) |
| Packaging | 6 Pk. |
| Quantity | 15 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction