Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Verax Biomedical™ Platelet PGDprime™ Test
Click to view available options
Quantity:
100 Tests
20 Tests
Description
- Easy to perform qualitative test
- Detects both aerobes and anaerobes
- Observed specificity: 100% / LCL: 99.9%
- 150 μL sample size
- No centrifugation or precision pipetting required
- Includes reagents and disposables required to perform the test
- Ready to use liquid reagents in dropper bottles
- Colored line in read window indicates a reactive result
- Built in procedural control verifies assay validity
- FDA cleared for use with: Leukoreduced apheresis platelets in plasma or PAS-C, polls of up to 6 pre-storage or post-storage pooled WBD platelets and individual WBD platelets
- CLIA Complexitiy: Moderate
- Ready to use liquid reagents in dropper bottles
- Colored line in read window indicates a reactive result
Specifications
Specifications
| CE Marker | No |
| Packaging Type | Box |
| Quantity | 100 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction