Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 234 HPPE Cut-Resistant Gloves, Foam Nitrile Coating

Catalog No. 19020486 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-020-486 Small
19-020-487 Medium
19-020-488 Large
19-020-489 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-020-486 Supplier SHOWA™ Supplier No. 234S

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Designed for use in 'knife hand' applications in food processing, these flexible cut protection gloves combine foam nitrile with a HPPE liner for protection in both dry and greasy environments.

  • 15-gauge seamless knit spandex liner with HPPE
  • Foamed flat dipped coating with sponge finish for secure grip
  • Contact Heat level 2
  • FDA and EU Food Contact approved
  • Lightweight, supple liner with breathable open back design
  • Foam nitrile coating protects against oils, hydrocarbons, grease and abrasions
  • Cooling HPPE properties and breathable back of hand reduces perspiration and keeps hands dry
  • Thin and lightweight for enhanced dexterity and longer use
  • Ergonomically designed to replicate the hand’s natural curves
  • Launderable for re-use, leads to less waste and cost efficiency
  • Seamless knit designed to prevent irritation

Specifications

Certifications/Compliance ANSI/ISEA 105-16: Level A4 (minimum 1500g)
Certifications/Compliance (Europe) EN 388:2016 Cut Level D
Construction Seamless Knit
Coating Material Sponge Nitrile
Color Gray, Black
Cut Resistance Rating ANSI A4
Latex Free No
Material HPPE, Spandex
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4X42D
For Use With (Application) HVAC, maintenance, mechanical and engineering, steel fabrication, washing and cleaning, component assembly
Liner Material HPPE
Model 234S
Series 234 Series
Yarn Gauge 15
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.