Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab Poly Scientific Special Stain Kit Component, Glutaraldehyde 2.5% In Phosphate Buffer pH 7.2

Click to view available options
:
8 oz.
Description
- A StatLab - Poly Scientific Special Stain Kit Component
- Used as a fixative in Electron Microscopy
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Colorless |
| Quantity | 8 oz. |
| Grade | Laboratory |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction