Learn More
Thermo Scientific™ Oxoid™ 24 LEB Complete
Description
Selectively enrichment of Listeria monocytogenes and other Listeria spp. from food and environmental samples with Thermo Scientific™ 24 Listeria Enrichment Broth (24 LEB) (Dehydrated). The medium provides optimal growing conditions for all Listeria species for PCR applications. It is specifically designed for use with PCR systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria.
Specifications
Specifications
| Certifications/Compliance | Adheres to ISO 11290-1:1997 standards. |
| Content And Storage | Room Temperature |
| Country of Origin | United Kingdom |
| Media Type | Dehydrated Culture Media |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Selective Enrichment of Listeria sp From Food and Environmental Samples |
| Packaging Type | Tub |
| Quantity | 10 kg |
| Species | Listeria |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.