Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

25,27-Dimethoxy-26-(N-trifluoroacetyl)carbamoyloxy-p-tert-butylcalix[4]arene, 95%, Thermo Scientific™

Catalog No. AC327482500
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 325776-12-3
Molecular Formula C49H60F3NO5
Molecular Weight (g/mol) 800.016
MDL Number MFCD03791046
InChI Key IZDRWLWARFNTMG-UHFFFAOYSA-N
PubChem CID 102601053
SMILES CC(C)(C)C1=CC2=CC(=C1)CC3=CC(=CC(=C3OC)CC4=C(C(=CC(=C4)C(C)(C)C)CC5=C(C(=CC(=C5)C(C)(C)C)C2)OC)OC(=O)NC(=O)C(F)(F)F)C(C)(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 815.98
Percent Purity ≥94%
Physical Form Granular Crystalline Powder
Chemical Name or Material 25, 27-Dimethoxy-26-(N-trifluoroacetyl)carbamoyloxy-p-tert-butylcalix[4]arene, 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.