Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ 25 mm Pendulum Bobs
SCI_ED

Catalog No. S41506 Shop All Eisco Products
Change view
Click to view available options
Material:
Aluminum
Brass
Copper
Iron
Iron, Copper, Aluminum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S41506 Aluminum
S27659 Iron
S41504 Brass
S41505 Copper
S41507 Iron, Copper, Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S41506 Supplier Eisco™ Supplier No. PH0306E
Add to Cart
Add to Cart

Includes

Bob with Threaded Hook

25 mm pendulum bobs are important tools while studying the physics of pendulum motion including tension force, gravity force, centripetal force, potential energy, kinetic energy, and more. Designed for use with full pendulum set up, not included.

  • Spherical pendulum bobs with hook,
  • 25mm in diameter
  • Hooks are threaded and removable
  • Useful for a variety physics experiments
  • Works well with Eisco pendulum clamp S96186

Warranty and Services

1 year limited manufacturer warranty

Specifications

Product Type Sphere
Material Aluminum
For Use With (Application) Pendulum Motion
Includes Bob with threaded hook
Quantity 1
Height (Metric) 2.5 cm
Weight (Metric) 20 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.