Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
25-OH Vitamin D3 Antibody-antigen Complex Antibody (RM428), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326039AF647
Description
25-OH Vitamin D3 Antibody-antigen Complex Monoclonal antibody specifically detects 25-OH Vitamin D3 Antibody-antigen Complex in Nonspecies specific samples. It is validated for ELISASpecifications
| 25-OH Vitamin D3 Antibody-antigen Complex | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| 25-OH Vitamin D3 Antibody-antigen Complex | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| RM428 | |
| ELISA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction