Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
25-OH Vitamin D3 Antibody-antigen Complex Antibody (RM428), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326039DL594
Description
25-OH Vitamin D3 Antibody-antigen Complex Monoclonal antibody specifically detects 25-OH Vitamin D3 Antibody-antigen Complex in Nonspecies specific samples. It is validated for ELISASpecifications
25-OH Vitamin D3 Antibody-antigen Complex | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
25-OH Vitamin D3 Antibody-antigen Complex | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
ELISA | |
RM428 | |
ELISA | |
Rabbit | |
Protein A purified | |
RUO | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction