Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 25-OH Vitamin D3 Recombinant Rabbit Monoclonal Antibody (RM428), Biotin
GREENER_CHOICE

Catalog No. PIMA544547
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544547 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA544547 Supplier Invitrogen™ Supplier No. MA544547
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody reacts to human CK18 (Cytokeratin-18).

25-OH Vitamin D3 (calcifediol) is a prohormone produced by the liver by hydroxylation of vitamin D3 and is used to measure vitamin D status and detect itamin D deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 25-OH Vitamin D3
Applications ELISA
Classification Recombinant Monoclonal
Clone RM428
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene Alias 25-hydroxy vitamin D; 25-hydroxyvitamin D; 25-OH Vitamin D
Host Species Rabbit
Immunogen 25-OH Vitamin D3 Antibody-antigen Complex.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.