Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 257 Aramid Knit Cut-Resistant Glove Gloves, Foam Nitrile Coating

Catalog No. 19020482 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
6, Small
7, Medium
8, Large
9, X-Large
10, 2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-020-482 7, Medium
19-020-481 6, Small
19-020-483 8, Large
19-020-484 9, X-Large
19-020-485 10, 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-020-482 Supplier SHOWA™ Supplier No. 25707/M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Soft and flexible gloves engineered with stainless steel and aramid knit to withstand the highest level of cuts and lacerations. ANSI Cut Level A7 is ideal for automotive, mechanical, engineering and various other industries.

  • 13-gauge plated-knit spandex liner with aramid and stainless steel
  • Foamed flat dipped coating with sponge finish for secure grip
  • Contact Heat level 2
  • Lightweight, flexible liner with elastic knitted wrist
  • Seamless knit and breathable open back design
  • Soft and flexible glove engineered with stainless steel and aramid to withstand the highest level of cuts and lacerations
  • Foam nitrile coating protects palm and fingers from abrasions, snags and punctures, while offering optimum grip in both dry and oily applications
  • Plated-knit liner avoids scratchy fibres touching the skin, ensuring a comfortable glove for long lasting wear
  • Lightweight, breathable liner with open back design reduces sweat and keeps hands dry
  • Ergonomically designed to replicate the hand’s natural curves
  • Launderable for re-use, leads to less waste and cost efficiency

Specifications

Certifications/Compliance ANSI/ISEA 105-16: Level A7 (minimum 4000g)
Certifications/Compliance (Europe) EN 388:2016: Cut Level F
Coating Material Sponge Nitrile
Color Yellow, Black
Cut Resistance Rating ANSI A7
Material Aramid Fiber, Spandex, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size 7, Medium
EN388 Standard EN 388:4X42F
For Use With (Application) Automotive, mechanical, engineering, metal stamping, glass, window manufacturing, component assembly
Liner Material Stainless Steel
Model 257-07
Series 257 Series
Yarn Gauge 13
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.