Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 27-Hydroxycholesterol
GREENER_CHOICESDP

Catalog No. 390710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-071-0 Supplier Tocris Bioscience™ Supplier No. 3907/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

LXR agonist; also ER partial agonist

27-Hydroxycholesterol is a liver X receptor (LXR) agonist. Also estrogen receptor partial agonist. Suppresses β-estradiol -mediated breast cancer cell proliferation in vitro. Also increases α-synuclein levels and induces apoptosis in human neuroblastoma cells in vitro. Endogenous cholesterol metabolite.

Specifications

Chemical Name or Material (3β,25R)-Cholest-5-ene-3,26-diol
CAS 20380-11-4
Synonym 27 OHC, 27 HC
Quantity 10 mg
Target LXR-like Receptor Agonists
Molecular Formula C27H46O2
Formula Weight 402.65
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.