Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ 293fectin™ Transfection Reagent

Catalog No. p-7180307
Encompass_Preferred
Click to view available options
Quantity:
1 mL
15 mL
10 x 15 mL

Includes

Each unit contains one vial of reagent.

293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells.

293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells. This reagent is optimized for transfecting suspension 293 human embryonic kidney cells (e.g., FreeStyle™ 293-F cells, Cat. No. R790-07) in defined, serum-free FreeStyle™ 293 Expression Medium (12338-018), and is intended for use with the FreeStyle™ 293 Expression System (K9000-01).

293fectin™ Transfection Reagent provides the following advantages:

  • Demonstrates high transfection efficiency in suspension 293 cells and is also suitable for transfecting adherent 293 cells
  • Suspension FreeStyle™ 293-F cells may be transfected in FreeStyle™ 293 Expression Medium; no medium change is required
  • Add 293fectin™ reagent complexes directly to cells in culture medium
  • No need to remove complexes or change or add medium following transfection

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each unit contains one vial of reagent. Store at 4°C.
293fectin™ is guaranteed stable for 6 months when stored properly.
Cell Type Established Cell Lines
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line 293fectin™
Product Type Transfection Reagent
Quantity 1 mL
Serum Compatible Yes
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.