Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2A Peptide Antibody (3H4), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259627AF700
Description
2A Peptide Monoclonal specifically detects 2A Peptide in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
2A Peptide | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
This 2A peptide antibody is specific for T2A and P2A tagged proteins. Specificity to F2A and E2A proteins has not been tested. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunocytochemistry, Immunofluorescence | |
3H4 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
2A cleavage peptide, 2A peptide, 2A peptide tag, 2A sequence, Anti- MDV 2A-peptide, anti-2A peptide, anti-2A sequence, anti-2A-like peptide, antibody for 2A-containing recombinant proteins, anti-P2A tagged proteins, anti-P2A peptide, anti-T2A tagged proteins, anti-T2A peptide, anti-Thosea asigna virus 2A, FMDV 2A-peptide peptide 2A sequences, self-cleaving'2A peptide2a pept, Thosea asigna virus 2A | |
KLH-coupled synthetic peptide corresponding to 2A peptide of Thosea asigna virus (CGDVEENPG) | |
0.1 mL | |
Primary | |
Non-species specific, Other | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction