Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2B4/CD244/SLAMF4 Antibody Pair (HRP), Novus Biologicals™

Rabbit Monoclonal Antibody
$875.53
Specifications
| Antigen | 2B4/CD244/SLAMF4 |
|---|---|
| Dilution | Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Host Species | Rabbit |
Description
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for ELISA, Sandwich ELISA.Specifications
| 2B4/CD244/SLAMF4 | |
| Monoclonal | |
| Rabbit | |
| 51744 | |
| Storage of components varies. See protocol for specific instructions. |
| Sandwich ELISA | |
| HRP | |
| Myeloid derived Suppressor Cell, Phospho Specific | |
| CD244 | |
| This solid phase sandwich ELISA utilizes a monoclonal antibody specific for 2B4/CD244/SLAMF4. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title