Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2B4/CD244/SLAMF4 Antibody Pair (HRP), R&D Systems™
SDP

Catalog No. NB003007 Shop All Novus Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003007 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003007 Supplier R&D Systems Supplier No. NBP27884515PLATES
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Antigen 2B4/CD244/SLAMF4
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Dilution Sandwich ELISA
Gene Alias CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Gene Symbols CD244
Host Species Rabbit
Quantity 15 Plates
Research Discipline Myeloid derived Suppressor Cell, Phospho Specific
Gene ID (Entrez) 51744
Test Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for 2B4/CD244/SLAMF4.
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.