Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2B4/CD244/SLAMF4 Antibody (146506), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB10392F
Description
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Flow Cytometry.Specifications
2B4/CD244/SLAMF4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CD244 | |
Mouse myeloma cell line NS0-derived partial recombinant human 2B4/CD244/SLAMF4 protein (amino acids 22-221) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Flow Cytometry | |
146506 | |
Flow Cytometry | |
CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
Mouse | |
Protein A or G purified | |
Myeloid derived Suppressor Cell, Phospho Specific | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction