Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2B4/CD244/SLAMF4, p Tyr271 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP146104
Description
2B4/CD244/SLAMF4 Polyclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
2B4/CD244/SLAMF4 (p Tyr271) | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody recognizes and epitope within the internal region. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000 | |
NP_057466.1 | |
CD244 | |
Peptide with sequence C-EFLTIpYEDVKD corresponding to internal region according to NP_057466.1. | |
0.1 mg | |
Phospho Specific | |
51744 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction