Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2B4/CD244/SLAMF4 Rabbit anti-Human, Clone: 20, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | 2B4/CD244/SLAMF4 |
---|---|
Clone | 20 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for ELISA.Specifications
2B4/CD244/SLAMF4 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
Recombinant Human 2B4/CD244/SLAMF4 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
20 | |
ELISA | |
Unconjugated | |
Rabbit | |
Myeloid derived Suppressor Cell, Phospho Specific | |
0.2 um filtered solution in PBS | |
51744 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title