Learn More
Description
2D Tubes with Screw Caps come preassembled and are made of virgin polypropylene materials for the highest level of safety against heavy metal ions and hazardous materials. Their internal thread provides optimal sealing for storage at ultralow temperatures, and silicone seals allow for secure air travel. Their internal thread provides optimal sealing for storage at ultralow temperatures, and silicone seals allow for secure air travel. The one-piece process allows the code to be integrated into the tube body.
The 1 mL tubes are available with a unique 2D Data-Matrix code. Due to the in-mold labeling technique used to manufacture the sample storage tubes, the white code surface cannot be separated from the transparent tube.
- Code is firm and non-shedding, resistant to acid, alkali, organic solvents and mechanical friction
- Produced in ISO Level 8 (Class 100,000 cleanroom) and automated production line to ensure DNase/RNase- and pyrogen-free
- Original coding, digital co-distribution technology and pure black and white ultra-high contrast Data-Matrix code ensure reliability of code reading
- Tubes with internal thread can be sealed with 96-format TPE caps
- Transportation safety in accordance with IATA requirements
Specifications
Specifications
| Certifications/Compliance | ISO13485:2016 and EU CE Certification (IVDD Directive) |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 1 mL |
| Bottom Shape | Flat |
| Material | Polypropylene (Medical Grade) |
| Thread Style | Internal |
| For Use With (Equipment) | 2D Barcode 96-well Rack |
| Quantity | 960/Pk. |
| Temperature Range (Metric) | -196°C to 121°C |
| Sterility | Sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

