Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ 2mL Ampoules (50 Pack)

Catalog No. 10486138 Shop All Osmonics Products
Change view
Click to view available options
Broth Type:
Cetrimide Broth
Enterococcus Broth
HPC Broth
HPC Broth with TTC
KF-Streptococcus Broth
M-Endo Coliform Broth
M-FC Broth
M-FC Broth with Rosolic Acid
M-Green Select Broth
M-Green Yeast and Mold Broth
M-TGE Total Count Liquid Media
MI-Broth Liquid Media
MRS Broth
Orange Serum Liquid Media
PRY Broth
Pseudomonas Broth
Total Count Broth with TTC
Detects:
Bacteria
Bacterial inhibition (low pH)
Enterococci counts in 48 hours
Fecal coliforms at 44.5°C
Fecal streptococci in water
Fungi, yeast and mold counts
Heterotrophs in water, media
Lactobacilli growth
Microbe spoilage-drinks/water
Pseudomonas Aeruginosa
aciduric microorganisms
coliform
presence of coliform bacteria
yeast and mold
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Broth Type Detects
10-486-138 M-Green Yeast and Mold Broth yeast and mold
10-486-139 M-TGE Total Count Liquid Media Bacteria
10-486-140 M-Endo Coliform Broth coliform
10-486-141 Orange Serum Liquid Media aciduric microorganisms
10-486-142 PRY Broth Microbe spoilage-drinks/water
10-486-145 MRS Broth Lactobacilli growth
10-486-146 Total Count Broth with TTC Bacteria
10-486-147 M-FC Broth with Rosolic Acid Bacterial inhibition (low pH)
10-486-148 M-Green Select Broth Fungi, yeast and mold counts
10-486-149 Pseudomonas Broth Pseudomonas Aeruginosa
10-486-150 Enterococcus Broth Enterococci counts in 48 hours
10-486-152 M-FC Broth Fecal coliforms at 44.5°C
10-486-153 KF-Streptococcus Broth Fecal streptococci in water
10-486-154 Cetrimide Broth Pseudomonas Aeruginosa
10-486-155 HPC Broth with TTC Heterotrophs in water, media
10-486-157 HPC Broth Heterotrophs in water, media
10-486-158 MI-Broth Liquid Media presence of coliform bacteria
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 10-486-138 Supplier GVS™ Supplier No. 10496101
Add to Cart
Add to Cart

Ready-to-use 2mL ampoule is a liquid culture growth media that can save time and money by shortening prep time and minimizing possible contamination. The 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish

  • Selective enrichment medium for use in the qualitative procedures for the isolation of Group BStreptococcus (GBS) from clinical specimens.
  • Selective growth promotion of GBS and inhibition of gram-negative organisms, such as Escherichia coli and Proteus mirabilis.
  • For in Vitro Diagnostic Use.
  • Premium medical grade plastic components to improve product shelf life.
  • For optimum performance, store at 2-25°C.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Appearance Green
Broth Type M-Green Yeast and Mold Broth
Certifications/Compliance Batch-specific quality certificate in each pack
Color Green
Content And Storage 2°C to 8°C
Detects yeast and mold
Format Ampule
Manufacturing Origin United States
Media Type M-Green Yeast and Mold Broth
Product Type Prepared Microbiology Tubes
Sterility Sterile
Target Organism Class Fungi
Test Type Fungal Testing
Fill Volume 2 to 2.4 mL
Formulation Yeast extract-9.0 gm,Dextrose (anhydrous)-50.0 gm,Pancreatic Digest of Casein-5.0 gm,Peptic Digest of Animal Tissue-5.0 gm,Magnesium Sulfate-2.1 gm,Potassium Phosphate-2.0 gmDiastase-0.05 gmThiamine-0.05 gmBromocresol Green-0.026 gm
For Use With (Application) Detection of Fungi in Beverages
Includes 50 Ampules, Labeled Foil Bag
Incubation Time 48 hr.
Material Polypropylene
No. per Pack 50
Packaging Type Ampoules
Quantity 2 mL
Dimensions (Dia. x H) 22 x 57.9 mm
Height (Metric) 57.9 mm
Diameter (Metric) 22 mm
Protein Source Yeast extract, Casein, Animal Tissue
Manufacturing Quality ISO 9001
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.