Learn More
Description
Features and Benefits
- Pre-Sterilized and Ready to Use
- Time Saving, 2 mL Portions, Reduce Risk of Cross-Contamination.
- 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish
- Optimal media stability, sterility, and reproducibility
- Less time-consuming, higher productivity
- Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability
Applications
- Microbiological analysis.
- Detection of fungi in routine analysis of beverages using membrane filter technique.
- For testing soft drinks and other beverages for growth of yeast and mold
Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits. This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.
See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.
Specifications
Specifications
| Broth Type | PRY Broth |
| Certifications/Compliance | Batch-specific quality certificate in each pack |
| Color | Pale Yellow |
| Content And Storage | 2°C to 8°C |
| Additive | Glacial Acetic Acid |
| Detects | Microbe spoilage-drinks/water |
| Format | Ampule, Liquid |
| Manufacturing Origin | United States |
| Media Type | PRY Broth |
| Product Type | Prepared Microbiology Tubes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.