Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ 2mL Ampoules (50 Pack)

Catalog No. 10486155 Shop All Osmonics Products
Change view
Click to view available options
Broth Type:
Cetrimide Broth
Enterococcus Broth
HPC Broth
HPC Broth with TTC
KF-Streptococcus Broth
M-Endo Coliform Broth
M-FC Broth
M-FC Broth with Rosolic Acid
M-Green Select Broth
M-Green Yeast and Mold Broth
M-TGE Total Count Liquid Media
MI-Broth Liquid Media
MRS Broth
Orange Serum Liquid Media
PRY Broth
Pseudomonas Broth
Total Count Broth with TTC
Detects:
Bacteria
Bacterial inhibition (low pH)
Enterococci counts in 48 hours
Fecal coliforms at 44.5°C
Fecal streptococci in water
Fungi, yeast and mold counts
Heterotrophs in water, media
Lactobacilli growth
Microbe spoilage-drinks/water
Pseudomonas Aeruginosa
aciduric microorganisms
coliform
presence of coliform bacteria
yeast and mold
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Broth Type Detects
10-486-155 HPC Broth with TTC Heterotrophs in water, media
10-486-138 M-Green Yeast and Mold Broth yeast and mold
10-486-139 M-TGE Total Count Liquid Media Bacteria
10-486-140 M-Endo Coliform Broth coliform
10-486-141 Orange Serum Liquid Media aciduric microorganisms
10-486-142 PRY Broth Microbe spoilage-drinks/water
10-486-145 MRS Broth Lactobacilli growth
10-486-146 Total Count Broth with TTC Bacteria
10-486-147 M-FC Broth with Rosolic Acid Bacterial inhibition (low pH)
10-486-148 M-Green Select Broth Fungi, yeast and mold counts
10-486-149 Pseudomonas Broth Pseudomonas Aeruginosa
10-486-150 Enterococcus Broth Enterococci counts in 48 hours
10-486-152 M-FC Broth Fecal coliforms at 44.5°C
10-486-153 KF-Streptococcus Broth Fecal streptococci in water
10-486-154 Cetrimide Broth Pseudomonas Aeruginosa
10-486-157 HPC Broth Heterotrophs in water, media
10-486-158 MI-Broth Liquid Media presence of coliform bacteria
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 10-486-155 Supplier GVS™ Supplier No. 10496151
Add to Cart
Add to Cart

Ready-to-use 2mL ampoule is a liquid culture growth media that can save time and money by shortening prep time and minimizing possible contamination. The 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish

Features and Benefits

  • Pre-Sterilized and Ready to Use
  • Time Saving, 2 mL Portions, Reduce Risk of Cross-Contamination.
  • 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish
  • Optimal media stability, sterility, and reproducibility
  • Less time-consuming, higher productivity
  • Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability

Applications

  • Microbiological analysis.
  • Detection of fungi in routine analysis of beverages using membrane filter technique.
  • For testing soft drinks and other beverages for growth of yeast and mold

Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits. This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Broth Type HPC Broth with TTC
Certifications/Compliance Batch-specific quality certificate in each pack
Color Pale Yellow
Content And Storage 2°C to 8°C
Additive TTC
Detects Heterotrophs in water, media
Format Ampule
Manufacturing Origin United States
Media Type HPC with TTC Indicator Broth
Product Type Prepared Microbiology Tubes
Sterility Sterile
Target Organism Class Bacteria
Test Type Standard Plate Count for Microorganism
Fill Volume 2 to 2.4 mL
Formulation Peptone: 30 gm, Gelatin: 15 gm, Glycerol: 15 ml,TTC, 1% solution: 10 ml
For Use With (Application) Heterotrophic Plate Count, Microbiological Analysis of Potable Water, Dairy Products, Swimming Pools
Includes 50 Ampules, Labeled Foil Bag
Incubation Conditions 35°C
Incubation Time 24 to 48 hr.
Material Polypropylene
No. per Pack 50
Packaging Type Ampoules
Quantity 2 mL
Dimensions (Dia. x H) 22 x 57.9 mm
Height (Metric) 57.9 mm
Diameter (Metric) 22 mm
Manufacturing Quality ISO 9001
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.