Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,3R)-1-Carboxy-4,5-dichloro-2,3-dihydroxycyclohexa-4,6-diene, 95%, Thermo Scientific™

Catalog No. AC271652500
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205587-32-2
Molecular Formula C7H6Cl2O4
Molecular Weight (g/mol) 225.021
MDL Number MFCD00142966
InChI Key VCMXBESUKVYUFQ-RITPCOANSA-N
Synonym 2r,3r-1-carboxy-4,5-dichloro-2,3-dihydroxycyclohex-4,6-diene, 5r,6r-3,4-dichloro-5,6-dihydroxycyclohexa-1,3-dienecarboxylic acid, 5r,6r-3,4-dichloro-5,6-dihydroxycyclohexa-1,3-diene-1-carboxylic acid, 1,3-cyclohexadiene-1-carboxylicacid, 3,4-dichloro-5,6-dihydroxy-, 5r,6r, 2r,3r-1-carboxy-4,5-dichloro-2,3-dihydroxycyanohexa-4,6-diene, 5r,6r-3,4-dichloro-5,6-dihydroxy-1,3-cyclohexadiene-1-carboxylic acid
PubChem CID 1268191
IUPAC Name (5R,6R)-3,4-dichloro-5,6-dihydroxycyclohexa-1,3-diene-1-carboxylic acid
SMILES C1=C(C(C(C(=C1Cl)Cl)O)O)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 225.03
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material (2R,3R)-1-Carboxy-4,5-dichloro-2,3-dihydroxycyclohexa-4,6-diene, 95%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.