Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,3R)-(-)-2,3-Butanediol, 98+%, 99+% ee

Catalog No. p-3757119
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 24347-58-8 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24347-58-8
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00064267
InChI Key OWBTYPJTUOEWEK-UHFFFAOYNA-N
Synonym 2r,3r-butane-2,3-diol, r,r-2,3-butanediol, 2r,3r---2,3-butanediol, r,r-butane-2,3-diol, 2r,3r-2,3-butanediol, r,r-2,3-butylene glycol, r,r---butane-2,3-diol, r,r---2,3-butylene glycol, unii-or02b2286a, r,r---2,3-butanediol
PubChem CID 225936
ChEBI CHEBI:16982
IUPAC Name (2R,3R)-butane-2,3-diol
SMILES CC(O)C(C)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 16.0°C
Color Colorless
Density 0.9800g/mL
Boiling Point 77.3°C to 77.4°C (10.0 mmHg)
Flash Point 85°C
Enantiomeric Excess 99.0% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4309 to 1.4329
Linear Formula CH3CH(OH)CH(OH)CH3
Quantity 1 g
Beilstein 01, I, 250
Fieser 16,52
Specific Gravity 0.98
Merck Index 15, 1571
Specific Rotation − 13.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and diisopropyl ether
Formula Weight 90.12
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material (2R, 3R)-(-)-2, 3-Butanediol, ≥99% ee
Show More Show Less
EINECSNumber 246-186-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.