Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, is used as a novel protected amino acid for the preparation of bestatin and analogs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, is used as a novel protected amino acid for the preparation of bestatin and analogs.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, is used as a novel protected amino acid for the preparation of bestatin and analogs.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Beilstein | 5909790 |
Solubility Information | Insoluble in water. Soluble in organic solvents. |
IUPAC Name | 3-{[(tert-butoxy)carbonyl]amino}-2-hydroxy-4-phenylbutanoic acid |
Formula Weight | 295.34 |
Percent Purity | 97% |
Quantity | 1 g |
Chemical Name or Material | (2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction