Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-(2S,8aR)-(Camphorylsulfonyl)oxaziridine, 98+%, Thermo Scientific™

Catalog No. AC294590010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104372-31-8
Molecular Formula C10H15NO3S
Molecular Weight (g/mol) 229.29
MDL Number MFCD00075428
InChI Key GBBJBUGPGFNISJ-UHFFFAOYNA-N
Synonym 1r---10-camphorsulfonyl oxaziridine, --2s,8ar-camphorylsulfonyl oxaziridine
PubChem CID 98044902
IUPAC Name 11,11-dimethyl-5-oxa-3λ⁶-thia-4-azatetracyclo[6.2.1.0¹,⁶.0⁴,⁶]undecane-3,3-dione
SMILES CC1(C)C2CCC11CS(=O)(=O)N3OC13C2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 168°C to 174°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 1 g
Specific Rotation -44° to -48° (20°C, 589nm) (c=2, chloroform)
Formula Weight 229.3
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material (-)-(2S, 8aR)-(Camphorylsulfonyl)oxaziridine, 99+%, (99+% e.e.)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.