Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1,2,3,6-Tetrahydro-4-pyridyl)indole, 95%, Thermo Scientific™

Catalog No. AAH6382103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6382103 1 g
AAH63821MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6382103 Supplier Thermo Scientific Chemicals Supplier No. H6382103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65347-55-9
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD03695471
InChI Key CIRSPTXGPFAXRE-UHFFFAOYSA-N
Synonym 3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, chembl27811, 3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 3-4-1,2,5,6-tetrahydropyridyl indole, thpindole, d02mvr, 3-1,2,3,6-tetrahydropyridin-4-yl indole, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indol, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indole
PubChem CID 2761023
IUPAC Name 3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
SMILES C1CNCC=C1C2=CNC3=CC=CC=C32

Specifications

Quantity 1 g
Beilstein 958099
Formula Weight 198.26
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(1,2,3,6-Tetrahydro-4-pyridyl)indole
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.