Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. p-7034061
Click to view available options
Quantity:
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257651-17-4
Molecular Formula C19H29BN2O5
Molecular Weight (g/mol) 376.26
MDL Number MFCD16294500
InChI Key IYTPRTWBNLJZLK-UHFFFAOYSA-N
Synonym 3-2-boc-amino acetamido benzeneboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl-carbamic acid tert-butyl ester, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamoyl methyl carbamate, amtb154, 3-2-boc-amino acetamido phenylboronic acid pinacol ester, 3-2-tert-butoxycarbonylamino acetamido phenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl carbamic acid tert-butyl ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylcarbamoyl methyl carbamic acid tert-butyl ester, tert-butyl 2-oxo-2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anilino ethyl carbamate
PubChem CID 57415693
IUPAC Name tert-butyl N-[2-oxo-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anilino]ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 5 g
Formula Weight 376.26
Percent Purity 97%
Chemical Name or Material 3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.