Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Ethylhexyl)thiophene 97.0+%, TCI America™
SDP

Catalog No. E09731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09731G 1 g
E0973200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09731G Supplier TCI America Supplier No. E09731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 121134-38-1
Molecular Formula C12H20S
Molecular Weight (g/mol) 196.352
MDL Number MFCD02260336
InChI Key HWMQCIZYXLAJKM-UHFFFAOYSA-N
PubChem CID 14419543
IUPAC Name 3-(2-ethylhexyl)thiophene
SMILES CCCCC(CC)CC1=CSC=C1

Specifications

Color Yellow
Boiling Point 30°C
Quantity 1 g
Formula Weight 196.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(2-Ethylhexyl)thiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.