Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Thienyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. T30551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30551G 1 g
T3055200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30551G Supplier TCI America Supplier No. T30551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21298-53-3
Molecular Formula C9H7NS
Molecular Weight (g/mol) 161.222
MDL Number MFCD01860052
InChI Key ZHBLIWDUZHFSJW-UHFFFAOYSA-N
Synonym 3-2-thienyl pyridine, 3-thiophen-2-yl pyridine, 2-3-pyridyl thiophene, 3-thiophen-2-yl-pyridine, pyridine,3-2-thienyl, 3-thien-2-ylpyridine, acmc-1clkc, 3-2-thienyl-pyridine, 3-thien-2-yl pyridine, 3-thiophene-2-yl-pyridine
PubChem CID 595145
IUPAC Name 3-thiophen-2-ylpyridine
SMILES C1=CC(=CN=C1)C2=CC=CS2

Specifications

Color Yellow
Boiling Point 100°C
Quantity 1 g
Formula Weight 161.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(2-Thienyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.