Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,3-Trifluoro-2,2-dimethylpropionic acid, 97%

Catalog No. AAH2605303
Click to view available options
Quantity:
250 mg
1 g

CAS: 889940-13-0 | C5H7F3O2 | 156.104 g/mol

3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 889940-13-0
Molecular Formula C5H7F3O2
Molecular Weight (g/mol) 156.104
MDL Number MFCD08445819
InChI Key VMFSJVUPIXOCFO-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-2,2-dimethylpropionic acid, 2-trifluoromethyl-2-methylpropanoic acid, 3,3,3-trifluoro-2,2-dimethyl-propionic acid, 2-trifluoromethyl-isobutyric acid, 3,3,3-trifluoro-2,2-dimethyl-propanoic acid
PubChem CID 19867367
IUPAC Name 3,3,3-trifluoro-2,2-dimethylpropanoic acid
SMILES CC(C)(C(=O)O)C(F)(F)F

Specifications

Melting Point 66°C to 73°C
Boiling Point 76°C to 77°C (14 mmHg)
Quantity 1 g
UN Number UN3261
Formula Weight 156.12
Percent Purity 97%
Chemical Name or Material 3,3,3-Trifluoro-2,2-dimethylpropionic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.