Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexene, 97%, Thermo Scientific™

Catalog No. AC312380050
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19430-93-4
Molecular Formula C6H3F9
Molecular Weight (g/mol) 246.076
MDL Number MFCD00042338
InChI Key GVEUEBXMTMZVSD-UHFFFAOYSA-N
Synonym perfluorobutyl ethylene, 3,3,4,4,5,5,6,6,6-nonafluoro-1-hexene, perfluorobutylethylene, 1h,1h,2h-perfluoro-1-hexene, 1h,1h,2h-perfluorohexene, perfluorobutyl ethene, 1-hexene, 3,3,4,4,5,5,6,6,6-nonafluoro, 3,3,4,4,5,5,6,6,6-nonafluorohexene, perfluoro-n-butyl ethylene, 1h,1h,2h-perfluorohex-1-ene
PubChem CID 88054
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohex-1-ene
SMILES C=CC(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.41g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula CF3(CF)3CH=CH
Quantity 5 g
Formula Weight 246.08
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexene, 97%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.