Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,4,4-Tetrafluoropyrrolidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T23425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23425G 5 g
T2342100MG 100 mg
T23421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T23425G Supplier TCI America Supplier No. T23425G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1810-13-5
Molecular Formula C4H6ClF4N
Molecular Weight (g/mol) 179.54
MDL Number MFCD06200835
InChI Key KPTMFLKIYHWQPB-UHFFFAOYSA-N
PubChem CID 22645920
IUPAC Name 3,3,4,4-tetrafluoropyrrolidine hydrochloride
SMILES Cl.FC1(F)CNCC1(F)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 179.54
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,4,4-Tetrafluoropyrrolidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.