Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetramethylbenzidine DihydroChloride, 98%, PROMO, Thermo Scientific Chemicals

Catalog No. J6033206PM
Click to view available options
Quantity:
5 g

Applications

3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.

Solubility

Soluble in DMSO (approx.1.7 mg/ml), and 1:300 DMSO:PBS (pH 7.2) (0.03 mg/mL).

Notes

Moisture and light sensitive. Store away from oxidizing agents, heat, light, acids, water/ moisture.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207738-08-7
Molecular Formula C16H22Cl2N2
Molecular Weight (g/mol) 313.27 g/mol
MDL Number MFCD00150104
InChI Key NYNRGZULARUZCC-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine dihydrochloride dihydrate, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride dihydrate, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, pubchem9044, tetramethylbenzidine dihydrate dihydrochloride
PubChem CID 19836601
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrate;dihydrochloride
SMILES Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1

Specifications

Color White
Quantity 5 g
Sensitivity Light and Moisture Sensitive
Solubility Information Soluble in DMSO, and 1:300 DMSO:PBS.
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA Yes
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.