Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetramethylbenzidine, MP Biomedicals™

Catalog No. 0219027701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-190-27701 1 g
02190277-1 100 mg
02-190-27705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-190-27701 Supplier MP Biomedicals, Inc Supplier No. 0219027701
Add to Cart
Add to Cart

3,3',5,5'-Tetramethylbenzidine is suitable as peroxidase substrate for use in ELISA procedures.

  • The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620-650 nm.
  • The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm.
  • A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases.
  • It is useful in gel staining procedure for low levels of home-associated peroxidases and for enzyme immunoassay of horseradish peroxidase.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.