Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,5,5-Tetramethylcyclohexanone, 98%

Catalog No. AC138390100
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle

CAS: 14376-79-5 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14376-79-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001640
InChI Key OQJMHUOCLRCSED-UHFFFAOYSA-N
Synonym 3,3,5,5-tetramethylcyclohexanone, cyclohexanone, 3,3,5,5-tetramethyl, 3,3,5,5-tetramethyl cyclohexanone, cyclohexanone,3,3,5,5-tetramethyl, acmc-1c4rg, 3,5,5-tetramethylcyclohexanone, cyclohexanone,3,5,5-tetramethyl, 3,3,5,5-tetramethyl-cyclohexanone, 3,3,5,5-tetramethyl-1-cyclohexanone, 3,3,5,5-tetramethyl-cyclohexan-1-one
PubChem CID 84399
IUPAC Name 3,3,5,5-tetramethylcyclohexan-1-one
SMILES CC1(CC(=O)CC(C1)(C)C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 11°C to 12°C
Color Colorless to Yellow
Density 0.8954g/mL
Boiling Point 200°C to 203°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4510 to 1.453
Linear Formula (CH3)4C6H6(=O)
Quantity 10 mL
Beilstein 07, IV, 89
Specific Gravity 0.8954
Formula Weight 154.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3,3,5,5-Tetramethylcyclohexanone
Show More Show Less
EINECSNumber 238-350-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.