Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine, TCI America™
SDP

Catalog No. B49071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49071G 1 g
B4907200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49071G Supplier TCI America Supplier No. B49071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1395881-58-9
Molecular Formula C36H20N2O4
Molecular Weight (g/mol) 544.566
InChI Key QKKFYYGKAXHDOK-UHFFFAOYSA-N
Synonym HN-D2
PubChem CID 89981752
SMILES C1=CC=C2C(=C1)N3C4=C(C=C(C=C4)C5=CC6=C(C=C5)N7C8=CC=CC=C8OC9=C7C(=CC=C9)O6)OC1=CC=CC(=C13)O2

Specifications

Melting Point 340°C
Color Yellow
Quantity 1 g
Formula Weight 544.57
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.