Learn More
Description
This product is sold in units of 359μg, equivalent to 0.5μmol.
Specifications
Specifications
| Chemical Name or Material | 3′,3′-Cyclic guanosine monophosphate-adenosine monophosphate sodium salt |
| CAS | 2407516-83-8 |
| Synonym | endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membr |
| Quantity | 359 μg |
| Target | STING/TMEM173 |
| Molecular Formula | C20H22N10Na2O13P2 |
| Formula Weight | 718.37 |
| Purity | >98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
