Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,3'-Diallylbisphenol A diacetate, epoxy curative, EC-392
Click to view available options
Quantity:
25 g
100 g
Description
It is used in epoxy systems or in combination with acrylates, methacrylates, and maleimides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Recommended for use in epoxy systems or in combination with acrylates, methacrylates, and maleimides

Specifications
Specifications
Color | Amber |
Physical Form | Amber liquid |
Quantity | 100 g |
Formula Weight | 392.5 |
Chemical Name or Material | 3,3'-Diallylbisphenol A diacetate, Epoxy curative, EC-392 |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction