Alfa Aesar™ 3,3'-Diaminobenzidine, 99%

Manufacturer: Alfa Aesar™ H5400006

Quantity 5g

 View more versions of this product

Catalog No. AAH5400006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 91-95-2
    Molecular Formula C12H14N4
    Formula Weight 214.27
    MDL Number MFCD00007725
    Synonym DAB; 3,3',4,4'-Tetraaminobiphenyl
    Chemical Name or Material 3,3'-Diaminobenzidine

    Specifications

    Formula Weight 214.27
    Health Hazard 2 GHS H Statement
    H350-H341
    May cause cancer.
    Suspected of causing genetic defects.
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 175° to 177°C
    Quantity 5g
    TSCA TSCA
    Molecular Formula C12H14N4
    MDL Number MFCD00007725
    CAS 91-95-2
    Synonym DAB; 3,3',4,4'-Tetraaminobiphenyl
    Health Hazard 1 Danger
    Recommended Storage 2° to 8°C
    RTECS Number DV8750000
    Chemical Name or Material 3,3'-Diaminobenzidine
    Assay 99%

    Description

    Applications
    3,3 ft.-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.

    Solubility
    Soluble in water (partly).

    Notes
    Light sensitive. Incompatible with oxidizing agents and heat.

    GHS H Statement
    H350-H341
    May cause cancer.
    Suspected of causing genetic defects.

    2° to 8°C

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit