Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dichloro-4,4'-diisocyanatobiphenyl 95.0+%, TCI America™
SDP

Catalog No. D15415G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5331-87-3
Molecular Formula C14H6Cl2N2O2
Molecular Weight (g/mol) 305.114
MDL Number MFCD00059591
InChI Key JITXMLLVGWGFGV-UHFFFAOYSA-N
PubChem CID 220082
IUPAC Name 2-chloro-4-(3-chloro-4-isocyanatophenyl)-1-isocyanatobenzene
SMILES C1=CC(=C(C=C1C2=CC(=C(C=C2)N=C=O)Cl)Cl)N=C=O

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Synonym 3,3′C-Dichlorobiphenyl-4,4′C-diyl Diisocyanate
Formula Weight 305.11
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dichloro-4,4′-diisocyanatobiphenyl
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.