Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diethylthiatricarbocyanine Iodide 98.0+%, TCI America™
SDP

Catalog No. D47731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47731G 1 g
D47735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47731G Supplier TCI America Supplier No. D47731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3071-70-3
Molecular Formula C25H25IN2S2
Molecular Weight (g/mol) 544.51
MDL Number MFCD00041994,MFCD00041994
InChI Key OYVFJKVYVDYPFV-UHFFFAOYSA-M
Synonym disc2 7, 3,3'-diethylthiatricarbocyanine iodide, 3-ethyl-2-7-3-ethyl-2 3h-benzothiazolylidene-1,3,5-heptatrienyl benzothiazolium iodide
PubChem CID 91972095
IUPAC Name 3-ethyl-2-[(5E)-7-[(2Z)-3-ethyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]hepta-1,3,5-trien-1-yl]-1,3-benzothiazol-3-ium iodide
SMILES [I-].CCN1\C(SC2=CC=CC=C12)=C\C=C\C=CC=CC1=[N+](CC)C2=CC=CC=C2S1

Specifications

Melting Point 211°C
Quantity 1 g
Formula Weight 544.51
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diethylthiatricarbocyanine Iodide
RTECSNumber DL7045000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.