Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diindolylmethane, 96%, Thermo Scientific™

Catalog No. p-7077781
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1968-05-4
Molecular Formula C17H14N2
Molecular Weight (g/mol) 246.31
ChEBI CHEBI:50182

Specifications

Melting Point 164.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Quantity 5 g
Synonym 3,3'-diindolylmethane, di 1h-indol-3-yl methane, diindolylmethane, 3,3'-methylenediindole, 3,3'-methylenebis-1h-indole, 1h-indole, 3,3'-methylenebis, dim, 3-diindolyl methane, arundine, 3,3'-methylenebis 1h-indole
InChI Key VFTRKSBEFQDZKX-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=CN2)CC3=CNC4=CC=CC=C43
IUPAC Name 3-(1H-indol-3-ylmethyl)-1H-indole
PubChem CID 3071
Formula Weight 246.31
Percent Purity 96%
Chemical Name or Material 3,3'-Diindolylmethane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.